Learn More
Sino Biological Anti-CD23 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10261MM20A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD23/FCER2 (rh CD23/FCER2; Catalog#10261-H07H; P06734-1; Asp48-Ser321) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-3817