Learn More
Sino Biological Anti-CD23 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10261MM20F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD23/FCER2 (rh CD23/FCER2; Catalog#10261-H07H; P06734-1; Asp48-Ser321) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3821