Learn More
Sino Biological Anti-CD23 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10261T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD23 / FCER2 (rh CD23 / FCER2; Catalog#10261-H07H; P06734-1; Asp48-Ser321). CD23 / FCER2 specific IgG was purified by Human CD23 / FCER2 affinity chromatography.
Catalog No. 50-172-3832