Learn More
Sino Biological Recombinant Human Noggin/NOG Protein (hFc Tag)
Catalog No. 501614474
Encompass_Preferred
A DNA sequence encoding the human Noggin precursor (NP_005441.1) (Met 1-Cys 232) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4474 Supplier Sino Biological Supplier No. 10267H02H100
Add to Cart
Add to Cart
A DNA sequence encoding the human Noggin precursor (NP_005441.1) (Met 1-Cys 232) was fused with the Fc region of human IgG1 at the C-terminus.