Learn More
Sino Biological Recombinant Human Noggin/NOG Protein (His Tag)
Supplier:  Sino Biological 10267H08H1
Encompass_Preferred
A DNA sequence encoding the human Noggin precursor (NP_005441.1) (Met 1-Cys 232) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4476