Learn More
Sino Biological Recombinant Human OMGP Protein (aa 1-416, His Tag), HPLC-verified
Supplier:  Sino Biological 10269H08H100
Encompass_Preferred
A DNA sequence encoding the mature form of human OMGP (P23515-1) (Met 1-Pro 416) without the pro peptide was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4485