Learn More
Sino Biological Anti-OMGP Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10269MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OMGP (rh OMGP; Catalog#10269-H08H; P23515-1; Met1-Pro416). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3882