Learn More
Sino Biological Recombinant Human Lymphotoxin alpha/TNF beta Protein
Supplier:  Sino Biological 10270HNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of human TNFβ (P01374) (Leu 35-Leu 205) was expressed and purified, with an initial Met at the N-terminus.
Catalog No. 50-161-4489