Learn More
Sino Biological Anti-Osteoprotegerin/TNFRSF11B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10271RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Osteoprotegerin / TNFRSF11B (rh TNFRSF11B; Catalog#10271-H08H; NP_002537.3; Met1-Leu401). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3921