Learn More
Sino Biological Anti-Prolactin Receptor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10278MM13100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Prolactin Receptor / PRLR (rh Prolactin Receptor / PRLR; Catalog#10278-H08H; NP_000940.1; Met1-Asp234). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3965