Learn More
Sino Biological Recombinant Anti-Prolactin Receptor Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10278R204100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PRLR (rh PRLR; Catalog#10278-H08H; NP_000940.1; Met 1-Asp 234).
Catalog No. 50-172-3969