Learn More
Sino Biological Recombinant Anti-Prolactin Receptor Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10278R204P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Prolactin Receptor / PRLR (rh Prolactin Receptor / PRLR; Catalog#10278-H08H; NP_000940.1; Met1-Asp234) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-3974