Learn More
Sino Biological Anti-Prolactin Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10278RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PRLR (rh PRLR; Catalog#10278-H08H; NP_000940.1; Met 1-Asp 234). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3981