Learn More
Sino Biological Anti-AXL Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10279MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AXL (rh AXL; Catalog#10279-H08H; AAB20305.1; Met 1-Pro 449). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3993