Learn More
Sino Biological Anti-AXL Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10279MM12F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AXL (rh AXL; Catalog#10279-H08H; AAB20305.1; Met1-Pro449) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3999