Learn More
Sino Biological Anti-AXL Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10279T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AXL (rh AXL; Catalog#10279-H08H; AAB20305.1; Met1-Pro449). AXL specific IgG was purified by Human AXL affinity chromatography.
Catalog No. 50-172-4020