Learn More
Sino Biological Recombinant Human Pin1 Protein (His Tag)
Supplier:  Sino Biological 10282H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human PIN1 (Q13526-1) (Met 1-Glu 163) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-4517