Learn More
Sino Biological Anti-Prostasin/PRSS8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10287RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Prostasin / PRSS8 (rh PRSS8; Catalog#10287-H08H; NP_002764.1; Ala30-Arg322). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4060