Learn More
Sino Biological Recombinant Human Pleiotrophin/PTN Protein
Supplier:  Sino Biological 10288HNAB1
Encompass_Preferred
A DNA sequence encoding the mature form of human PTN (NP_002816.1) (Met1-Asp168) was expressed.
Catalog No. 50-161-4526