Learn More
Sino Biological Anti-Galectin-3/LGALS3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10289MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Galectin-3 / LGALS3 (rh Galectin-3 / LGALS3; Catalog#10289-HNAE; P17931; Met1-Ile250). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4069