Learn More
Sino Biological Anti-Galectin-3/LGALS3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10289RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LGALS3 / Galectin-3 (rh LGALS3 / Galectin-3; Catalog#10289-HNAE; P17931; Met1-Ile250). LGALS3 / Galectin-3 specific IgG was purified by Human LGALS3 / Galectin-3 affinity chromatography.
Catalog No. 50-172-4085