Learn More
Sino Biological Anti-Galectin-1/LGALS1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10290MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Galectin-1 (rh Galectin-1; Catalog#10290-HNAE; NP_002296.1; Met1-Asp135). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4091