Learn More
Sino Biological Recombinant Anti-Galectin-1/LGALS1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10290R008100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Galectin-1 (rh Galectin-1; Catalog#10290-HNAE; NP_002296.1; Met1-Asp135).
Catalog No. 50-172-4103