Learn More
Sino Biological Recombinant Human PD-L2/B7-DC Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10292H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PD-L2 (NP_079515.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4542