Learn More
Sino Biological Recombinant Anti-PD-L2/B7-DC Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10292R002F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human B7-DC / PD-L2 / CD273 / PDCD1LG2 (rh B7-DC / PD-L2 / CD273 / PDCD1LG2; Catalog#10292-H08H; NP_079515.2; Met1-Pro219) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4127