Learn More
Sino Biological Anti-COQ9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102948T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human COQ9, and purified by antigen affinity chromatography.
Catalog No. 50-172-4141