Learn More
Sino Biological Anti-SerpinD1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10295MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinD1 / HC2 (rh SerpinD1 / HC2; Catalog#10295-H08H; NP_000176.2; Met 1-Ser 499). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4150