Learn More
Sino Biological Anti-PAI-1/Serpin E1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10296RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinE1 (rh SerpinE1; Catalog#10296-H08H; NP_000593.1; Met 1-Pro 402). SerpinE1 specific IgG was purified by human SerpinE1 affinity chromatography.
Catalog No. 50-172-4177