Learn More
Sino Biological Recombinant Anti-Alpha 2 Antiplasmin/SerpinF2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10297R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinF2 (rh SerpinF2; Catalog#10297-H08H; NP_000925.2; Met1-Lys491).
Catalog No. 50-172-4194