Learn More
Sino Biological Recombinant Human Mer Protein (aa 578-872)
Supplier:  Sino Biological 10298HNCB150
Encompass_Preferred
A DNA sequence encoding the human MERTK (Q12866) (Glu 578-Tyr 872) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-4562