Learn More
Sino Biological Anti-Mer Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10298MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MERTK/Mer (rh MERTK/Mer; Catalog#10298-HCCH; NP_006334.2; Met1-Ala499). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4211