Learn More
Enterprise Technology Solutions 103 Odor Counteractant Concentrate, Cherry
SDP
Supplier:  Enterprise Technology Solutions FRS1232WBCH
Encompass_Preferred
Water-soluble deodorant cleans, deodorizes, refreshes.Powerful detergents and potent perfumes counteract malodors and leave area smelling fresh and clean.Add 1-2 oz. per gallon of water or cleaning solution.Use in fabric shampoo, mop water, any water-based solution.Can be used in trigger sprayers, carpet extractors, laundry.
Catalog No. 50-211-0913