Learn More
Sino Biological Recombinant Human Galectin-8/LGALS8 Protein (GST Tag)
Supplier:  Sino Biological 10301H09E100
Encompass_Preferred
A DNA sequence encoding human LGALS8 (AAF19370.1) (Met 1-Trp 317) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-4565