Learn More
Sino Biological Anti-Galectin-8/LGALS8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10301RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LGALS8 / Galectin-8 (rh LGALS8 / Galectin-8; Catalog#10301-H09E; AAF19370.1; Met 1-Trp 317). LGALS8 / Galectin-8 specific IgG was purified by Human LGALS8 / Galectin-8 affinity chromatography.
Catalog No. 50-172-4247