Learn More
Sino Biological Recombinant Human Factor XI Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10302H08H50
Encompass_Preferred
A DNA sequence encoding the human F11 (NP_000119.1) precursor (Met 1-Val 625) with a carboxy-terminal polyhistidine tag was expressed.
Catalog No. 50-161-4571