Learn More
Sino Biological Anti-Factor XI Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10302T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Coagulation Factor XI/F11 (rh Coagulation Factor XI/F11; Catalog#10302-H08H; NP_000119.1; Met1-Val625). Coagulation Factor XI/F11 specific IgG was purified by Human Coagulation Factor XI/F11 affinity chromatography.
Catalog No. 50-172-4266