Learn More
Sino Biological Recombinant Anti-Serpin A1/Alpha-1-antitrypsin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10306R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinA1 (rh SerpinA1; Catalog#10306-H08H; NP_000286.3; Met1-Lys418).
Catalog No. 50-172-4289