Learn More
Sino Biological Anti-Serpin A1/Alpha-1-antitrypsin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10306RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinA1 (rh SerpinA1; Catalog#10306-H08H; NP_000286.3; Met 1-Lys 418). SerpinA1 specific IgG was purified by Human SerpinA1 affinity chromatography.
Catalog No. 50-172-4299