Learn More
Sino Biological Anti-SerpinA3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10307MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinA3 (rh SerpinA3; Catalog#10307-H08H; NP_001076.2; Met 1-Ala 423). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4308