Learn More
Sino Biological Anti-FAM46A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103072T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FAM46A, and purified by antigen affinity chromatography.
Catalog No. 50-172-4302