Learn More
Refrigiwear Inc. 1030R EXTREME SOCK SIZE L/XL EXTREME MERINO WOOL SOCK
SDP
Catalog No. NC3867057
Encompass_Preferred
You are always ready for whatever the job takes - even overtime. Make sure your feet are, too. With the Extreme Socks you don't just get warmth and - you get footwear that lasts as long as you do. Advanced construction means you get a cushioned sole with a breathable top that can take all the abuse of a full day - plus more. The entire Extreme Collection, now with head to toe protection, was designed for comfort, style, and the ultimate protection. When the cold warns you to stay at home, take it to the Extreme. 72% Merino wool / 26% acrylic / 2% spandex Arch support Hi-splice nylon reinforcements in toe and heel Welted top keeps socks fitting right Mesh ventilation on top of foot for maximum breathability  
Catalog No. NC3867057 Supplier Refrigiwear Inc. Supplier No. 1030RHVLLXL
You are always ready for whatever the job takes - even overtime. Make sure your feet are, too. With the Extreme Socks you don't just get warmth and - you get footwear that lasts as long as you do. Advanced construction means you get a cushioned sole with a breathable top that can take all the abuse of a full day - plus more. The entire Extreme Collection, now with head to toe protection, was designed for comfort, style, and the ultimate protection. When the cold warns you to stay at home, take it to the Extreme. 72% Merino wool / 26% acrylic / 2% spandex Arch support Hi-splice nylon reinforcements in toe and heel Welted top keeps socks fitting right Mesh ventilation on top of foot for maximum breathability  