Learn More
Sino Biological Recombinant Human ROBO2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10310H08H100
Encompass_Preferred
A DNA sequence encoding the human ROBO2 (Q9HCK4-1) (Met1-Pro859) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4578