Learn More
Sino Biological Anti-GFRAL Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103115T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GFRAL, and purified by antigen affinity chromatography.
Catalog No. 50-172-4351