Learn More
Sino Biological Anti-GLIPR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103118T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GLIPR1 / RTVP1, and purified by antigen affinity chromatography.
Catalog No. 50-172-4354