Learn More
Sino Biological Recombinant Anti-Fibronectin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10314R014100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fibronectin Fragment 2 (Catalog#10314-H08H; Ser 607-Pro 1265; NP_997647.1).
Catalog No. 50-172-4366