Learn More
Sino Biological Anti-Fibronectin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10314RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Fibronectin Fragment 2 (Catalog#10314-H08H; Ser 607-Pro 1265; NP_997647.1). Fibronectin Fragment 2 specific IgG was purified by human Fibronectin Fragment 2 affinity chromatography.
Catalog No. 50-172-4371