Learn More
Sino Biological Anti-Flt3 Ligand/FLT3LG Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10315MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FLT3L / Flt3 ligand (rh FLT3L / Flt3 ligand; Catalog#10315-H07B; P49771-1; Thr27-Pro185). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4380