Learn More
Sino Biological Anti-Flt3 Ligand/FLT3LG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10315RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FLT3L / Flt3 ligand (rh FLT3L / Flt3 ligand; Catalog#10315-H07B; P49771-1; Thr27-Pro185). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4385