Learn More
Sino Biological Anti-HOXA10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103169T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HOXA10, and purified by antigen affinity chromatography.
Catalog No. 50-172-4391