Learn More
Sino Biological Recombinant Human CDH12 Protein (His Tag)
Supplier:  Sino Biological 10317H08H100
Encompass_Preferred
A DNA sequence encoding the pro form of human CDH12 (NP_001304156.1) extracellular domain (Met 1-Ala 605) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4584